Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
CDCA5 is the regulator of sister chromatid cohesion in mitosis. It may act by regulating the ability of the cohesin complex to mediate sister chromatid cohesion, perhaps by altering the nature of the interaction of cohesin with the chromosomes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell division cycle-associated protein 5; p35; Sororin; sororin p35
Gene Aliases: 2610036L13Rik; AL024086; AW536684; C85404; CDCA5; RGD1560863; SORORIN
UniProt ID: (Human) Q96FF9, (Mouse) Q9CPY3
Entrez Gene ID: (Human) 113130, (Rat) 684771, (Mouse) 67849
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support