Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEKDGLCRA DQQYECVAEI GEGAYGKVFK ARDLKNGGRF VALKRVRVQT GEEGMPLSTI REVAVLRHLE TFEHPNVVRL FDVCTVSRTD RETKLTLVFE HVDQDLTTYL DKVPEPGVPT ETIKDMMFQL LRGLDFLHSH RVVHRDLKPQ NILVTSSGQI KLADFGLARI YSFQMALTSV VVTLWYRAPE VLLQSSYATP VDLWSVGCIF AEMFRRKPLF RGSSDVDQLG KILDVIGLPG EEDWPRDVAL PRQAFHSKSA QPIEKFVTDI DELGKDLLLK CLTFNPAKRI SAYSALSHPY FQDLERCKEN LDSHLPPSQN TSELNTA (1-326 aa encoded by BC027989)
CDK6 functions as a cell-cycle initiator protein. This protein is coexpressed and copurified with CyclinD1. They are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression. CDK6 is essential for cell proliferation within the dentate gyrus of the hippocampus and the cuventricular zone of the lateral ventricles. Mutations in the gene results in microcephaly 12, primary, autosomal recessive (MCPH12).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell division protein kinase 6; CR2 protein kinase; CRK2; Cyclin-dependent kinase 6; MGC59692; Serine/threonine-protein kinase PLSTIRE
Gene Aliases: 5830411I20; AI504062; CDK6; CDKN6; Crk2; MCPH12; PLSTIRE
UniProt ID: (Human) Q00534, (Mouse) Q64261
Entrez Gene ID: (Human) 1021, (Rat) 114483, (Mouse) 12571
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support