Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TEEEPDDKG DKNQQQQQGN NHTNGTGHPG NQDSSHTQGP PLKKVRVVPP TTTSGGLIMT SDYQRSNPHA AYPNPGPSTS QPQSSMGYSA TSQQPPQYSH QTHRY (360-464 aa encoded by BC069634)
CDK8 phosphorylates the carboxy-terminal domain of the largest subunit of RNA polymerase II. This protein is coexpressed and copurified with Cyclin C. CDK8 is part of a family of proteins designated as cdks are critical regulators of cell cycle progression. The prototype member of this family, p34/Cdc 2, and a related protein Cdk 2, function late in the cycle while Cdk 4 and Cdk 6 are critically involved in G1 to S progression. Because of analogy with yeast Cdk 8, it has been proposed that the human Cdk 8 Cyclin C may interact with RNA polymerase, thus influencing the transcription mechanism(s) that lead to cell division.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDK8 protein kinase; Cell division protein kinase 8; Cyclin-dependent kinase 8; Mediator complex subunit CDK8; Mediator of RNA polymerase II transcription subunit CDK8; MGC126074; MGC126075; Protein kinase K35
Gene Aliases: CDK8; K35
UniProt ID: (Human) P49336, (Mouse) Q8R3L8
Entrez Gene ID: (Human) 1024, (Mouse) 264064
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support