Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KLTIESTPFN VAEGKEVLLL AHNLPQNRIG YSWYKGERVD GNSLIVGYVI
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
Predicted homology: Guinea Pig: 79%; Human: 100%
CEA Cell Adhesion Molecule 6 (CEACAM6) belongs to the carcinoembryonic antigen (CEA) family whose members are glycosyl phosphatidyl inositol (GPI) anchored cell surface glycoproteins. Members of this family play a role in cell adhesion and are widely used as tumor markers in serum immunoassay determinations of carcinoma. CEACAM6 affects the sensitivity of tumor cells to adenovirus infection. The acts as a receptor for adherent-invasive E. coli adhesion to the surface of ileal epithelial cells in patients with Crohn's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carcinoembryonic antigen-related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific cross reacting antigen); CD66c; CEACAM-1; CEACAM-3; CEACAM-5; CEACAM-6; CGM1; Cluster of Differentiation 66c; Non-specific crossreacting antigen; Normal cross-reacting antigen; pan CEACAM
Gene Aliases: CD66c; CEACAM6; CEAL; NCA
UniProt ID: (Human) P40199
Entrez Gene ID: (Human) 4680
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support