Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive Samples: SH-SY5Y, U-87MG, Mouse brain, Rat brain
Immunogen sequence: ALLNNHSNLK PAPTVPSSPD ATPEPKGPGD GAEEDEAASG GPGGRGPWSC ENFNPLLVAG GVAVAAIALI LGVAFLVRKK
The protein encoded by this gene is a neuron-specific protein. The similar protein in pig enhances neuroblastoma cell differentiation in vitro and may be involved in neuronal differentiation in vivo. Multiple pseudogenes have been reported for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BM88 antigen; Cell cycle exit and neuronal differentiation protein 1
Gene Aliases: 1500001H12Rik; AI415214; BM88; C38; CEND1; RGD1309401
UniProt ID: (Human) Q8N111, (Rat) Q5FVI4, (Mouse) Q9JKC6
Entrez Gene ID: (Human) 51286, (Rat) 361675, (Mouse) 57754
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support