Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MGPRRRSRKP EAPRRRSPSP TPTPGPSRRG PSLGASSHQH SRRRQGWLKE IRKLQKSTHL LIRKLPFSRL AAEAFLVHLF EDAYLLTLHA GRVTLFPKDV QLARRIRGLE EGLG
Centromeres are the differentiated chromosomal domains that specify the mitotic behavior of chromosomes. CENPA encodes a centromere protein which contains a histone H3 related histone fold domain that is required for targeting to the centromere. CENPA is proposed to be a component of a modified nucleosome or nucleosome-like structure in which it replaces 1 or both copies of conventional histone H3 in the (H3-H4)2 tetrameric core of the nucleosome particle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CENP A; CENP-A; Centromere autoantigen A; Centromere protein A; centromere protein A, 17kDa; centromere-specific histone; Histone H3-like centromeric protein A
Gene Aliases: CenH3; CENP-A; CENPA
UniProt ID: (Human) P49450
Entrez Gene ID: (Human) 1058
Molecular Function: chromatin/chromatin-binding, or -regulatory protein histone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support