Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
ARAP1 (angiotensin II type 1 receptor-associated protein) is an adaptor protein with ArfGAP and RhoGAP activities, containing five PH domains and ankyrin repeat. This adaptor seems to serve as a link between phosphoinositide- Arf-, and Rho-mediated cell signaling. ARAP1 supports the plasma membrane recycling of angiotensin II type 1 receptor (AT1) and is important also for cell-specific trafficking of pro-death receptor TRAIL-R1 (DR4) to the plasma membrane, thus promoting the TRAIL-induced apoptosis in certain cell types. ARAP1 also affects cell spreading.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 1; ARF-GAP, RHO-GAP, ankyrin repeat, and pleckstrin homology domains-containing protein 1; centaurin, delta 2; Centaurin-delta-2; Cnt-d2
Gene Aliases: ARAP1; CENTD2; KIAA0782
UniProt ID: (Human) Q96P48
Entrez Gene ID: (Human) 116985
Molecular Function:
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support