Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAESSSESDH FRCRDRLSPW AARSTHRGTR SLPTVEVTEK VNTITSTLQD TSRNLRQVDQ MLGRYREYSN GQAGAIEHLK ESLEQSIDQL RSQRLLRNSG GRSISVTSLS ASDLDGGTGS ELHHFPPTSP LKDYGDPQGI KRMRSRTGVR FVQETDDMTQ LHGFHQSLRD LSSEQIRLGD DFNRELSRRS RSDAETKRAL EELTEKLNEA QKQEVVSDRV ERRLQELERE MRTERELVER RQDQLGLMSL QLQEALKKQE AKADEHEGAI KNKLRQTETE KNQLEQELEL SRRLLNQSEG SRETLLHQVE ELRTQLTKAE GDRKGLQHQV SQISKQQSNY QDEQGEDWRF RRGVEREKQD LEKQMSDLRV QLNFSAMASE LEEVKRCMER KDKERAHLAS QVEHINRKCL LNLVQDLDCK DNEILTYSLQ SPLHV
CEP128 gene ontology annotations related to this gene include protein localization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: centrosomal protein 128kDa; Centrosomal protein of 128 kDa; Cep128; lens epithelium-derived protein
Gene Aliases: C14orf145; C14orf61; CEP128; LEDP/132
UniProt ID: (Human) Q6ZU80
Entrez Gene ID: (Human) 145508
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support