Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
CEP128 gene ontology annotations related to this gene include protein localization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: centrosomal protein 128kDa; Centrosomal protein of 128 kDa; Cep128; lens epithelium-derived protein
Gene Aliases: C14orf145; C14orf61; CEP128; LEDP/132
UniProt ID: (Human) Q6ZU80
Entrez Gene ID: (Human) 145508
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support