Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LTVDWSRARD ELMRKESQWQ MEQEFFKGYL KGEHGRLLSL WREVVTFRRH FLEMKSATDR DLMELKAEHV RLSGSLLTCC LRLTVGAQSR EPNGSGRMDG
Probably plays an important role in centrosome cohesion during interphase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 250 kDa centrosomal protein; C-Nap1; Centrosomal Nek2-associated protein 1; Centrosomal protein 2; centrosomal protein 250kDa; centrosome associated protein; Centrosome-associated protein CEP250; Cep250
Gene Aliases: C-NAP1; CEP2; CEP250; CNAP1
UniProt ID: (Human) Q9BV73
Entrez Gene ID: (Human) 11190
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support