Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CEP63 is a protein with six coiled-coil domains. The protein is localized to the centrosome, a non-membranous organelle that functions as the major microtubule-organizing center in animal cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: centrosomal protein 63kDa; Centrosomal protein of 63 kDa; centrosome protein CEP63; Cep63
Gene Aliases: CEP63; SCKL6
UniProt ID: (Human) Q96MT8
Entrez Gene ID: (Human) 80254
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support