Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQRGPASGDP REGPPRQRRE GCWGLHTCLI TAARLRQSGW EHDLGVCSGS TDDKLWKLAV CKALASLLLL KCQIPMLYID AKCLTQPGLG AVRRKLAIRG GGAGPGLPQD SSEAGPEVCT RGPDLSLSFL HTEFSIFIEH LVLLSQSSVN CVSDVCLLPR SHDGSLVSSA ARGLRRPEDS SRKAFLPRSP GHPSIVYYVL V
CERK converts ceramide to ceramide 1-phosphate (C1P), a sphingolipid metabolite. Both CERK and C1P have been implicated in various cellular processes, including proliferation, apoptosis, phagocytosis, and inflammation (Kim et al., 2006 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acylsphingosine kinase; Ceramide kinase; CERK antibody; hCERK; Lipid kinase 4; lipid kinase LK4; LK4
Gene Aliases: CERK; dA59H18.2; dA59H18.3; hCERK; KIAA1646; LK4
UniProt ID: (Human) Q8TCT0
Entrez Gene ID: (Human) 64781
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support