Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the carboxylesterase large family. The family members are responsible for the hydrolysis or transesterification of various xenobiotics, such as cocaine and heroin, and endogenous substrates with ester, thioester, or amide bonds. They may participate in fatty acyl and cholesterol ester metabolism, and may play a role in the blood-brain barrier system. This enzyme is the major liver enzyme and functions in liver drug clearance. Mutations of this gene cause carboxylesterase 1 deficiency. Three transcript variants encoding three different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACAT; acyl coenzyme A:cholesterol acyltransferase; Acyl-coenzyme A:cholesterol acyltransferase; Brain carboxylesterase hBr1; Carboxylesterase 1; carboxylesterase 1 (monocyte/macrophage serine esterase 1); carboxylesterase 2 (liver); CE-1; CEH; Cholesteryl ester hydrolase; Cocaine carboxylesterase; Egasyn; HMSE; human monocyte/macrophage serine esterase 1; Liver carboxylesterase 1; Methylumbelliferyl-acetate deacetylase 1; MGC117365; Monocyte/macrophage serine esterase; REH; Retinyl ester hydrolase; Serine esterase 1; TGH; triacylglycero; Triacylglycerol hydrolase
Gene Aliases: ACAT; CE-1; CEH; CES1; CES2; hCE-1; HMSE; HMSE1; PCE-1; REH; SES1; TGH
UniProt ID: (Human) P23141
Entrez Gene ID: (Human) 1066
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support