Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QREKHNGGRE EVTKVATQKH RQSPLNWTSS HFGEVTGSAE GWGPEEPLPY SRAFGEGASA RPRCCRN
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CFC-1; cripto; cryptic family 1; Cryptic family protein 1; Cryptic protein; CRYPTICFLJ77897; heterotaxy 2 (autosomal dominant); HTX2MGC133213
Gene Aliases: CFC1; CFC1B; CRYPTIC; DTGA2; HTX2
UniProt ID: (Human) P0CG37
Entrez Gene ID: (Human) 55997
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support