Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 14
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: CGRP.
This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-type CGRP; Calcitonin; Calcitonin gene-related peptide 1; Calcitonin gene-related peptide I; calcitonin/calcitonin-related polypeptide, alpha; CGRP-I; MGC126648; RAT calcitonin
Gene Aliases: CA; Cal1; CAL6; Calc; Calc1; Calca; calcitonin; CGRP; CGRP-1; CGRP1; Ct; Ctn; RATCAL6
UniProt ID: (Rat) P01256, (Mouse) Q99JA0
Entrez Gene ID: (Rat) 24241, (Mouse) 12310
Molecular Function:
peptide hormone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support