Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: LDNTNLEKFS DGAFLGVTTL KHVHLENNRL NQLPSNFPFD SLETLALTNN PWKCTCQLRG LRRWLEAKAS RPDATCASPA KFKGQHIRDT DAFRSCKFPT KRSKKAGRH
Chondroadherin is a cartilage matrix protein thought to mediate adhesion of isolated chondrocytes. The protein contains 11 leucine-rich repeats flanked by cysteine-rich regions. The chondroadherin messenger RNA is present in chondrocytes at all ages.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cartilage leucine-rich protein; Chondroadherin; chondroadherin proteoglycan
Gene Aliases: CHAD; SLRR4A
UniProt ID: (Human) O15335
Entrez Gene ID: (Human) 1101
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support