Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CHAT is known to be present exclusively in the cytoplasm of cholinergic neurons and anti-CHAT antibody has been considered as the best marker for cholinergic neurons.
Immunogen sequence: SDTHRALQL LHGGGYSKNG ANRWYDKSLQ FVVGRDGTCG VVCEHSPFDG IVLVQCTEHL LKHMTQSSRK LIRADSVSEL PAPRRLRWKC SPEIQGHLAS SAEKLQRIVK NLDFIVYKFD NYGKTFIKKQ KCSPDAFIQV ALQLAFYRLH RRLVPTYESA SIRRFQEGRV DNIRSATPEA LAFVRAVTDH KAAVPASEKL LLLKDAIRAQ TAYTVMAITG MAIDNHLLAL RELARAMCKE LPEMFMDETY LMSNRFVLST SQVPTTTEMF CCYGPVVPNG YGACYNPQPE TILFCISSFH SCKETSSSKF AKAVEESLID MRDLCSLLPP TESKPLATKE KATRPSQGHQ P (399-748 aa encoded by BC130617)
ChAT catalyzes the reversible synthesis of acetylcholine (ACh) from acetyl CoA and choline at cholinergic synapses.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetyl CoA; acetyl CoA:choline O-acetyltransferase; CHOACTase; choline acetylase; choline acetyltransferase; Choline O-acetyltransferase
Gene Aliases: B230380D24Rik; CHAT; CHOACTASE; CMS1A; CMS1A2; CMS6
UniProt ID: (Human) P28329
Entrez Gene ID: (Human) 1103, (Rat) 290567, (Mouse) 12647
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support