Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MQAALEVTA RYCGRELEQY GQCVAAKPES WQRDCHYLKM SIAQCTSSHP IIRQIRQACA QPFEAFEECL RQNEAAVGNC AEHMRRFLQC AEQVQPPRSP ATVEAQPLPA S (1-110 aa encoded by BC004498)
CHCHD5 (Coiled-Coil-Helix-Coiled-Coil-Helix Domain Containing 5) is a Protein Coding gene. Among its related pathways are Metabolism of proteins and Mitochondrial protein import.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil-helix-coiled-coil-helix domain-containing protein 5; mitochondrial intermembrane space cysteine motif protein of 14 kDa homolog
Gene Aliases: C2orf9; CHCHD5; MIC14
UniProt ID: (Human) Q9BSY4
Entrez Gene ID: (Human) 84269
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support