Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the CHD family of proteins which are characterized by the presence of chromo domains and SNF2-related helicase/ATPase domains. This protein is one of the components of a histone deacetylase complex referred to as the Mi-2/NuRD complex which participates in the remodeling of chromatin by deacetylating histones. Chromatin remodeling is essential for many processes including transcription. Autoantibodies against this protein are found in a subset of patients with dermatomyositis. Three alternatively spliced transcripts encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent helicase Chd3; CG9594-PA; Chd3-PA; Chromodomain-helicase-DNA-binding protein 3; MGC40857; Mi 2a; Mi2 ALPHA; Prp9 1
Gene Aliases: CG9594; CHD-3; CHD3; dCHD3; DmCHD3; Dmel\CG9594; Dmel_CG9594
UniProt ID: (Fruit fly) O16102
Entrez Gene ID: (Fruit fly) 40111
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support