Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control: 293T, A431, HeLa, HepG2, U87-MG, SK-N-SH, IMR32, SK-N-AS
Predicted Reactivity: Mouse (93%), Rat (94%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CHD5 (Chromodomain helicase DNA binding protein 5) is a member of the SNF2/RAD54 helicase family of chromatin remodeling and DNA-binding proteins (CDH proteins). Heavily expressed in both fetal and adult brain, CHD5 plays a role in nervous system development and acts as a tumor suppressor via the Arf/p53 pathway. CHD5, along with other chromo-domain proteins, forms remodeling complexes, such as NuRD, that promote normal neuroblast maturation and are thought to prevent overexpression of neuronal cells. Errors in these chromatin remodeling complexes can leave the cell in a perpetual state of growth, preventing differentiation and leading to tumor formation. Due to the importance of the CHD proteins in proper brain development, deletions in the gene encoding CHD5 are commonly found in neuroblastomas, suggesting that CHD5 deficiency may lead to malignant cell transformation and metastasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent helicase CHD5; CHD-5; CHD5; Chromodomain-helicase-DNA-binding protein 5
Gene Aliases: CHD-5; CHD5; KIAA0444
UniProt ID: (Human) Q8TDI0
Entrez Gene ID: (Human) 26038
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support