Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGNLFGRKK QSRVTEQDKA ILQLKQQRDK LRQYQKRIAQ QLERERALAR QLLRDGRKER AKLLLKKKRY QEQLLDRTEN QISSLEAMVQ SIEFTQIEMK VMEGLQFGNE CLNKMHQVMS IEEVERILDE TQEAVEYQRQ IDELLAGSFT QEDEDAILEE LSAITQEQIE LPEVPSEPLP EKIPENVPVK ARPRQAELVA AS (1-201 aa encoded by BC010108)
CHMP6 belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A, is required for both MVB formation and regulation of cell cycle progression (Tsang et al., 2006 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Charged multivesicular body protein 6; chromatin modifying protein 6; Chromatin-modifying protein 6; hVps20; novel SNF7 protein; Vacuolar protein sorting-associated protein 20; Vps20
Gene Aliases: 2400004G01Rik; CHMP6; RGD1565325; VPS20
UniProt ID: (Human) Q96FZ7, (Mouse) P0C0A3
Entrez Gene ID: (Human) 79643, (Mouse) 208092, (Rat) 287873
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support