Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CSIQVTYFP FDWQNCTMVF SSYSYDSSEV SLQTGLGPDG QGHQEIHIHE GTFIENGQWE IIHKPSRLIQ PPGDPRGGRE GQRQEVIFYL IIRRKPLFYL VNVIAPCILI TLLAIFVFYL PPDAGEKMGL SIFALLTLTV FLLLLADKVP ETSLSVPIII KYLMFTMVLV TFSVILSVVV LNLHHRSPHT HQMPLWVRQI FIHKLPLYLR LKRPKPERDL MPEPPHCSSP GSGWGRGTDE YFIRKPPSDF LFPKPNRFQP ELSAPDLRRF IDGPNRAVAL LPELREVVSS ISYIARQLQE QEDHDALKED WQFVAMVVDR LFLWTFIIFT SVGTLVIFLD ATYHLPPPDP FP (151-501 aa encoded by BC023553)
The muscle acetylcholine receptor is composed of five subunits: two alpha subunits and one beta, one gamma, and one delta subunit. This gene encodes the beta subunit of the acetylcholine receptor. The acetylcholine receptor changes conformation upon acetylcholine binding leading to the opening of an ion-conducting channel across the plasma membrane. Mutations in this gene are associated with slow-channel congenital myasthenic syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetylcholine receptor; Acetylcholine receptor beta; Acetylcholine receptor subunit beta; acetylcholine receptor, nicotinic, beta 1 (muscle); AChR beta; cholinergic receptor; cholinergic receptor nicotinic beta polypeptide 1; cholinergic receptor, nicotinic beta 1; cholinergic receptor, nicotinic, beta 1 (muscle); cholinergic receptor, nicotinic, beta polypeptide 1 (muscle); Chrnb 1; nAChRB1
Gene Aliases: Achr-2; ACHRB; Acrb; CHRNB; CHRNB1; CMS1D; CMS2A; CMS2C; RNACRB1; SCCMS
UniProt ID: (Human) P11230, (Rat) P25109, (Mouse) P09690
Entrez Gene ID: (Human) 1140, (Rat) 24261, (Mouse) 11443
Molecular Function:
ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support