Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HILILATTR SGSSFVGQLF NQHLDVFYLF EPLYHVQNTL IPRFTQGKSP ADRRVMLGAS RDLLRSLYDC DLYFLENYIK PPPVNHTTDR IFRRGASRVL CSRPVCDPPG PADLVLEEGD CVRKCGLLNL TVAAEACRER SHVAIKTVRV PEVNDLRALV EDPRLNLKVI QLVRDPRGIL ASRSETFRDT YRLWRLWYGT GRKPYNLDVT QLTTVCEDFS NSVSTGLMRP PWLKGKYMLV RYEDLARNPM KKTEEIYGFL GIPLDSHVAR WIQNNTRGDP TLGKHKYGTV RNSAATAEKW RFRLSYDIVA FAQNACQQVL AQLGYKIAAS EEELKNPSVS LVEERDFRPF S (62-411 aa encoded by BC022567)
Carbohydrate sulfotransferase 1 (CHST1), also referred to as KSGal6ST or KSST, is a keratan sulfate sulfotransferase. It is predominantly expressed in brain and skeletal muscle and localizes to the trans-Golgi network. CHST1 is responsible for mediating the sulfation of keratan in the cornea, which is important in maintaining corneal transparency. In particular, CHST1 catalyzes the transfer of sulfate groups from 3'-phosphoadenosine 5'-phosphosulfate to position six of internal or terminal galactose residues (preferentially the residues adjacent to sulfated GlcNAc) on growing keratan sulfate chains. CHST1 also contributes to the generation of L-selectin ligands. Mutations in the CHST1 gene may play a role in macular corneal dystrophy (MCD).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carbohydrate (chondroitin 6/keratan) sulfotransferase 1; carbohydrate (keratan sulfate Gal-6) sulfotransferase 1; Carbohydrate sulfotransferase 1; Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 1; GST-1; Keratan sulfate Gal-6 sulfotransferase; Keratan sulfotransferase; KS6ST; KSGal6ST; KSST; N-acetylglucosamine-6-O-sulfotransferase 1
Gene Aliases: 2610008E20Rik; AW125896; C6ST; CHST1; GST-1; Gst1; KS6ST; KSGAL6ST; KSST
UniProt ID: (Human) O43916, (Rat) Q5RJQ0, (Mouse) Q9EQC0
Entrez Gene ID: (Human) 8534, (Rat) 295934, (Mouse) 76969
Molecular Function: transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support