Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LDLLLLYKK HKGKKMTVPV RRHAYLQQTF SKIQFVEHEE LDAQELAKRI NQESGSLSFL SNSLKKLVPF QLPGSKSEHK EPKDKKINIL IPLSGRFDMF VRFMGNFEKT CLIPNQNVKL VVLLFNSDSN PDKAKQVELM RDYRIKYPKA DMQILPVSGE FSRALALEVG SSQFNNESLL FFCDVDLVFT TEFLQRCRAN TVLGQQIYFP IIFSQYDPKI VYSGKVPSDN HFAFTQKTGF WRNYGFGITC IYKGDLVRVG GFDVSIQGWG LEDVDLFNKV VQAGLKTFRS QEVGVVHVHH PVFCDPNLDP KQYKMCLGSK ASTYGSTQQL AEMWLEKNDP SYSKSSNNNG SVRTA (449-802 aa encoded by BC046247)
CHSY1 synthesizes chondroitin sulfate, a glycosaminoglycan expressed on the surface of most cells and in extracellular matrices. Glycosaminoglycan chains are covalently linked to a wide range of core protein families and regulate many biologic processes, including cell proliferation and recognition, extracellular matrix deposition, and morphogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carbohydrate (chondroitin) synthase 1; carbohydrate synthase 1; Chondroitin glucuronyltransferase 1; chondroitin glucuronyltransferase II; Chondroitin sulfate synthase 1; Chondroitin synthase 1; ChSy-1; glucuronosyl-N-acetylgalactos; Glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase 1; N-acetylgalactosaminyl-proteoglycan 3-beta-glucuronosyltransferase 1; N-acetylgalactosaminyltransferase 1; N-acetylgalactosaminyltransferase II
Gene Aliases: CHSY; ChSy-1; CHSY1; CSS1; KIAA0990; mKIAA0990; TPBS; UNQ756/PRO1487
UniProt ID: (Human) Q86X52, (Mouse) Q6ZQ11
Entrez Gene ID: (Human) 22856, (Mouse) 269941, (Rat) 292999
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support