Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: RQKEIWHLLK IACTQSSARS LVGSSLEGAV TPQTSAWLPP TSAEHDHSLS CVVTPQDGET SAQMIEENLN CLGHLSTIIH EANEEQGNSM MNLDWSWLTE
CHUK (IKK alpha) is a serine/threonine kinase involved in signal transduction and cellular communication. It belongs to the cytokine activated protein complex that is an inhibitor of the NF-kappa B complex. IKK alpha along with IKK beta and a regulatory protein NEMO (IKK gamma) phosphorylates IkB proteins which targets the later for ubiquitin mediated degradation. It is involved with several cellular processes with extensive studies on its role in inflammation and cell survival.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Conserved helix-loop-helix ubiquitous kinase; EC 2.7.11.10; FLJ40509; I-kappa-B kinase 1; I-kappa-B kinase alpha; I-kappa-B kinase-alpha; ikappaB kinase; IkB kinase alpha subunit; IKBKB; IKK-1; IKK-a kinase; IKK-B; IKK-beta; IKKB; Inhibitor of nuclear factor kappa-B kinase subunit alpha; kinase beta; MGC131801; NFKBIKA; Nuclear factor NF-kappa-B inhibitor kinase alpha; Nuclear factor NFkappaB inhibitor kinase alpha; TCF-16; Transcription factor 16
Gene Aliases: CHUK; IKBKA; IKK-alpha; IKK1; IKKA; NFKBIKA; TCF16
UniProt ID: (Human) O15111
Entrez Gene ID: (Human) 1147
Molecular Function:
non-receptor serine/threonine protein kinase
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support