Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ALVTATLGG EELEPAPSLP RPLPATVGVT QPYLDRLGYR RTDHDDPAFK RNGFRINLAK PRPGDPAEAN GPVYPWRSLR ECQGAPVTAS HFRFARVEAD KYEYNVVPFR EGTPASWTGD LLAWWPNPQE FRACFLKVKI QGPQEYMVRS HNAGGSHPRT RGQLYGLRDA RSVRDPERPG TSAACVEFKC SGMLFDQRQV DRTLVTIMPQ GSCRRVAVNG LLRDYLTRHP PPVPAEDPAA FSMLAPLDPL GHNYGVYTVT DQSPRLAKEI AIGRCFDGSS DGFSREMKAD AGTAVTFQCR EPPAGRPSLF QRLLESPATA LGDIRREMSE AAQAQARASG PLRTRRGRVR Q (304-653 aa encoded by BC034926)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cartilage intermediate layer protein 2; cartilage intermediate layer protein-like protein 2; CILP-2
Gene Aliases: 1110031K21Rik; AA407300; CILP2; CLIP-2
UniProt ID: (Human) Q8IUL8, (Mouse) D3Z7H8
Entrez Gene ID: (Human) 148113, (Mouse) 68709
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support