Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSLTSSSSVR VEWIAAVTIA AGTAAIGYLA YKRFYVKDHR NKAMINLHIQ KDNPKIVHAF DMEDLGDKAV YCRCWRSKKF PFCDGAHTKH NEETGDNVGP LIIKKKET
This gene encodes a protein with a CDGSH iron-sulfur domain and has been shown to bind a redox-active [2Fe-2S] cluster. The encoded protein has been localized to the outer membrane of mitochondria and is thought to play a role in regulation of oxidation. Genes encoding similar proteins are located on chromosomes 4 and 17, and a pseudogene of this gene is located on chromosome 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDGSH iron sulfur domain-containing protein 1; CDGSH iron-sulfur domain-containing protein 1; Cysteine transaminase CISD1; MGC14684; MitoNEET; zinc finger CDGSH-type domain 1; zinc finger, CDGSH-type domain 1
Gene Aliases: AU043990; AW743335; C10orf70; CISD1; D10Ertd214e; MDS029; MitoNEET; RGD1309529; ZCD1
UniProt ID: (Human) Q9NZ45, (Mouse) Q91WS0, (Rat) B0K020
Entrez Gene ID: (Human) 55847, (Mouse) 52637, (Rat) 294362
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support