Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This gene encodes a member of the CREB-binding protein/p300-interacting transactivator with Asp/Glu-rich C-terminal domain (CITED) family of proteins. The encoded protein, also known as melanocyte-specific gene 1, may function as a transcriptional coactivator and may play a role in pigmentation of melanocytes. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cbp/p300-interacting transactivator 1; Cbp/p300-interacting transactivator, with Glu/Asp rich carboxy-terminal domain, 1; Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 1; melanocyte-specific gene 1; Melanocyte-specific protein 1
Gene Aliases: CITED1; MSG1
UniProt ID: (Human) Q99966
Entrez Gene ID: (Human) 4435
Molecular Function:
transcription cofactor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support