Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.
Colonic and hepatic tumor over-expressed protein (ch-TOG) belongs to the TOG/XMAP215/Dis1 evolutionarily ancient family of microtubule-associated proteins with isoforms found in all major kingdoms of eukaryotes. This protein is required for the assembly and function of the meiotic or mitotic spindles during cell division and is localized to microtubule-organizing centers (MTOCs) during interphase or to the spindle poles and centrosomes during mitosis or cytokinesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ch-TOG; ch-TOGp; ckap5; colonic and hepatic tumor over-expressed gene protein; Colonic and hepatic tumor overexpressed gene protein; Cytoskeleton-associated protein 5
Gene Aliases: 3110043H24Rik; 4930432B04Rik; ch-TOG; CHTOG; CKAP5; D730027C18Rik; KIAA0097; mKIAA0097; MSPS; T25636; TOG; TOGp
UniProt ID: (Human) Q14008, (Mouse) A2AGT5
Entrez Gene ID: (Human) 9793, (Mouse) 75786, (Rat) 311191
Molecular Function: cytoskeletal protein microtubule or microtubule-binding cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support