Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody can recognize both CKB and CKM due to the high homology.
Immunogen sequence: MPFGNTHNK FKLNYKPEEE YPDLSKHNNH MAKVLTLELY KKLRDKETPS GFTVDDVIQT GVDNPGHPFI MTVGCVAGDE ESYEVFKELF DPIISDRHGG YKPTDKHKTD LNHENLKGGD DLDPNYVLSS RVRTGRSIKG YTLPPHCSRG ERRAVEKLSV EALNSLTGEF KGKYYPLKSM TEKEQQQLID DHFLFDKPVS PLLLASGMAR DWPDARGIWH NDNKSLLVWV NEEDHLRVIS MEKGGNMKEV FRRFCVGLQK IEEIFKKAGH PFMWNQHLGY VLTCPSNLGT GLRGGVHVKL AHLSKHPKFE EILTRLRLQK RGTGGVDTAA VGSVFDVSNA DRLGSSEVEQ VQLVVDGVKL MVEMEKKLEK GQSIDDMIPA QK (1-381 aa encoded by BC007462)
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-CK; Brain creatine kinase; CK MB; CPK-B; CPK-M; Creatine kinase B chain; Creatine kinase B-type; creatine kinase brain; creatine kinase brain-type; Creatine kinase M chain; Creatine kinase M-type; creatine kinase, brain; creatine kinase, muscle; creatine kinase-B; creatine kinase-M; Creatine phosphokinase B-type; Creatine phosphokinase M-type; epididymis luminal protein 211; epididymis secretory protein Li 29; M-CK
Gene Aliases: B-CK; BCK; Ck-3; Ck3; CKB; CKBB; CKM; CKMM; HEL-211; HEL-S-29; M-CK; MCK
UniProt ID: (Human) P12277, (Human) P06732, (Mouse) Q04447, (Mouse) P07310
Entrez Gene ID: (Human) 1152, (Human) 1158, (Mouse) 12709, (Mouse) 12715
Molecular Function:
amino acid kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support