Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEPRMESCL AQVLQKDVGK RLQVGQELID YFSDKQKSAD LEHDQTMLDK LVDGLATSWV NSSNYKVVLL GMDILSALVT RLQDRFKAQI GTVLPSLIDR LGDAKDSVRE QDQTLLLKIM DQAANPQYVW DRMLGGFKHK NFRTREGICL CLIATLNASG AQTLTLSKIV PHICNLLGDP NSQVRDAAIN SLVEIYRHVG ERVRADLSKK GLPQSRLNVI FTKFDEVQKS GNMIQSAND (1-238 aa encoded by BC112940 )
CLASPs, such as CLASP1, are nonmotor microtubule-associated proteins that interact with CLIPs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CLIP-associating protein 1; cytoplasmic linker associated protein 1; Cytoplasmic linker-associated protein 1; hOrbit1; multiple asters 1; Multiple asters homolog 1; Protein Orbit homolog 1
Gene Aliases: 1700030C23Rik; 5730583A19Rik; B130045P17Rik; CLASP1; CLASP1alpha; KIAA0622; MAST1
UniProt ID: (Human) Q7Z460, (Mouse) Q80TV8
Entrez Gene ID: (Human) 23332, (Mouse) 76707, (Rat) 304740
Molecular Function:
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support