Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DGLAKHPPPF SQPNSIPTAP SHKALRRSYS PSMLDYDTEN LNSEEIYSSL RGVTEAIEKF SFRSQEDLNE PIKRDGKKEC DIVSRDGGAA SPAT
CLASPs, such as CLASP1, are nonmotor microtubule-associated proteins that interact with CLIPs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CLIP-associating protein 1; Cytoplasmic linker-associated protein 1; hOrbit1; multiple asters 1; Multiple asters homolog 1; Protein Orbit homolog 1
Gene Aliases: CLASP1; KIAA0622; MAST1
UniProt ID: (Human) Q7Z460
Entrez Gene ID: (Human) 23332
Molecular Function:
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support