Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
This gene encodes a member of the calcium sensitive chloride conductance protein family. To date, all members of this gene family map to the same region on chromosome 1p31-p22 and share a high degree of homology in size, sequence, and predicted structure, but differ significantly in their tissue distributions. The encoded protein is expressed as a precursor protein that is processed into two cell-surface-associated subunits, although the site at which the precursor is cleaved has not been precisely determined. The encoded protein may be involved in mediating calcium-activated chloride conductance in the intestine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CaCC-1; Calcium-activated chloride channel family member 1; Calcium-activated chloride channel protein 1; Calcium-activated chloride channel regulator 1; calcium-dependent chloride channel-1; chloride channel regulator 1; chloride channel, calcium activated, family member 1; CLCA family member 1, chloride channel regulator; hCLCA1
Gene Aliases: CACC; CaCC-1; CACC1; CLCA1; CLCRG1; GOB5; hCaCC-1; hCLCA1
UniProt ID: (Human) A8K7I4
Entrez Gene ID: (Human) 1179
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support