Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
1 µg/mL of MA5-45554 was sufficient for detection of Clcn3 in 20 µg of rat brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 90kDa. Does not cross-react with Clcn4 or Clcn5 (based on KO validation results).
This antibody was formerly sold as clone S258-5.
Mediates the exchange of chloride ions against protons. Functions as antiporter and contributes to the acidification of the endosome and synaptic vesicle lumen, and may thereby affect vesicle trafficking and exocytosis. May play an important role in neuronal cell function through regulation of membrane excitability by protein kinase C. It could help neuronal cells to establish short-term memory.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chloride channel 3; Chloride channel protein 3; chloride channel, voltage-sensitive 3; Chloride transporter ClC-3; CIC3; ClC-3; CLCN 3; DKFZp564I0463; H(+)/Cl(-) exchange transporter 3; protein kinase C-regulated chloride channel
Gene Aliases: ClC-3; CLC3; CLCN3
UniProt ID: (Human) P51790, (Rat) P51792, (Mouse) P51791
Entrez Gene ID: (Human) 1182, (Rat) 84360, (Mouse) 12725
Molecular Function:
ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support