Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MGCRDVHAAT VLSFLCGIAS VAGLFAGTLL PNWRKLRLIT FNRNEKNLTV YTGLWVKCAR YDGSSDCLMY DTTWYSSVDQ LDLRVLQFAL PLSMLIAMGA LLLCLIGMCN TAFRSSVPNI KLAKCLVNSA GCHLVAGLLF FLAGTVSLSP SIWVIFYNIH LNKKFEPVFS FDYAVYVTIA SAGGLFMTSL ILFIWYCTCK SLPSPFWQPL YSHPPSMHTY SQPYSARSRL SAIEIDIPVV SHTT
The claudin superfamily consists of many structurally related proteins in humans. These proteins, which include claudin 1 through 18, are important structural and functional components of tight junctions in paracellular transport. Claudins are located in both epithelial and endothelial cells in all tight junction-bearing tissues. Claudins, which consist of four trans-membrane domains and two extracellular loops, make up tight junction strands.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Claudin-12
Gene Aliases: CLDN12
UniProt ID: (Human) D6W5Q4
Entrez Gene ID: (Human) 9069
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support