Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Rat anti Mouse CLEC2 antibody, clone 17D9 recognizes mouse C-type lectin domain family 1 member B, also known as CLEC2 or CLEC1B.
Natural killer cells express multiple calcium-dependent lectin-like receptors, such as CD94 and NKG2D, that interact with major histocompatibility complex class I molecules and either inhibit or activate cytotoxicity and cytokine secretion. CLEC2 is a C-type lectin-like receptor expressed in myeloid cells and NK cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-type lectin domain family 1 member B; C-type lectin-like receptor 2; CLEC-2
Gene Aliases: 1810061I13Rik; Clec-2; Clec1b; Clec2
UniProt ID: (Mouse) Q9JL99
Entrez Gene ID: (Mouse) 56760
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support