Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QYCTDMNATL LKIDNRNIVE YIKARTHLIR WVGLSRQKSN EVWKWEDGSV ISENMFEFLE DGKGNMNCAY FHNGKMHPTF CENKHYLMCE RKAGMTKVDQ LP
Natural killer cells express multiple calcium-dependent lectin-like receptors, such as CD94 and NKG2D, that interact with major histocompatibility complex class I molecules and either inhibit or activate cytotoxicity and cytokine secretion. CLEC2 is a C-type lectin-like receptor expressed in myeloid cells and NK cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-type lectin domain family 1 member B; C-type lectin domain family 1, member B; C-type lectin-like receptor 2; C-type lectin-like receptor-2; CLEC-2
Gene Aliases: 1810061I13Rik; CLEC1B; CLEC2; CLEC2B; PRO1384; QDED721; UNQ721/PRO1384
UniProt ID: (Human) Q9P126
Entrez Gene ID: (Human) 51266
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support