Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: NFMYSKTVKR LSKLREYQQY HPSLTCVMEG KDIEDWSCCP TPWTSFQSSC YFISTGMQSW TKSQKNCSVM GADLVVINTR EEQDFIIQNL KRNSSYFLGL SDPGGRRHWQ WVDQTPYNEN VTFWHSGEPN NLDERCAIIN FRSSEEWGWN DIHCHVPQKS ICKMKKIYI
This gene encodes a member of the C-type lectin/C-type lectin-like domain superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may play a role in dendritic cell function. Two transcript variants encoding distinct isoforms have been identified for this gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BDCA-2; Blood dendritic cell antigen 2; blood dendritic cell antigen 2 protein; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 11; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 7; C-type lectin domain family 4 member C; C-type lectin domain family 4, member C; C-type lectin domain family 4, member C, isoform 1; C-type lectin superfamily member 7; CD303; CD303 antigen; dendritic cell lectin b; Dendritic lectin
Gene Aliases: BDCA2; CD303; CLEC4C; CLECSF11; CLECSF7; DLEC; HECL; PRO34150; UNQ9361/PRO34150
UniProt ID: (Human) Q8WTT0
Entrez Gene ID: (Human) 170482
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support