Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
CLEC9A is a group V C-type lectin-like receptor (CTLR) that functions as an activation receptor and is expressed on myeloid lineage cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-type lectin domain family 9 member A; C-type lectin domain family 9, member A; CD370; HEEE9341
Gene Aliases: CD370; CLEC9A; DNGR-1; DNGR1; UNQ9341; UNQ9341/PRO34046
UniProt ID: (Human) Q6UXN8
Entrez Gene ID: (Human) 283420
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support