Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MALSMPLNG LKEEDKEPLI ELFVKAGSDG ESIGNCPFSQ RLFMILWLKG VVFSVTTVDL KRKPADLQNL APGTHPPFIT FNSEVKTDVN KIEEFLEEVL CPPKYLKLSP KHPESNTAGM DIFAKFSAYI KNSRPEANEA LERGLLKTLQ KLDEYLNSPL PDEIDENSME DIKFSTRKFL DGNEMTLADC NLLPKLHIVK VVAKKYRNFD IPKEMTGIWR YLTNAYSRDE FTNTCPSDKE VEIAYSDVAK RLTK (1-253 aa encoded by BC012444)
Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 4 (CLIC4) protein, encoded by the CLIC4 gene, is a member of the p64 family; the gene is expressed in many tissues and exhibits a intracellular vesicular pattern in Panc-1 cells (pancreatic cancer cells).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chloride intracellular channel 4 (mitochondrial); chloride intracellular channel 4 like; Chloride intracellular channel protein 4; DKFZP566G223; FLJ38640; Intracellular chloride ion channel protein p64H1; mitochondrial chloride intracellular channel 4
Gene Aliases: CLIC4; CLIC4L; H1; huH1; MTCLIC; p64H1
UniProt ID: (Human) Q9Y696, (Rat) Q9Z0W7
Entrez Gene ID: (Human) 25932, (Pig) 100622787, (Rat) 83718
Molecular Function:
ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support