Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user.
Immunogen sequence: FICPPKQGLF ASVSKISKAV DAPPSSVTST PRTPRMDFSR VTGKGRREHK GKKKTPSSPS LGSLQQRDGA KAEVGDQVLV AGQKQGIV
Functions as a cytoplasmic linker protein. Involved in TGN-endosome dynamics.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAP-Gly domain-containing linker protein 3; CLIP-170-related 59 kDa protein; Cytoplasmic linker protein 170-related 59 kDa protein; restin-like 1
Gene Aliases: CLIP3; CLIPR-59; CLIPR59; RSNL1
UniProt ID: (Human) Q96DZ5
Entrez Gene ID: (Human) 25999
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support