Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Could play a role in cell proliferation during neuronal differentiation and in protection against cell death.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ceroid-lipofuscinosis, neuronal 8 (epilepsy, progressive with mental retardation); motor neuron degeneration; Protein CLN8
Gene Aliases: C8orf61; CLN8; EPMR; mnd
UniProt ID: (Human) Q9UBY8, (Mouse) Q9QUK3, (Rat) Q6AYM9
Entrez Gene ID: (Human) 2055, (Mouse) 26889, (Rat) 306619
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support