Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
ATP-dependent specificity component of the Clp protease. It directs the protease to specific substrates. Can perform chaperone functions in the absence of clpP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent Clp protease ATP-binding subunit clpX-like, mitochondrial; caseinolytic peptidase X; caseinolytic protease X; ClpX caseinolytic peptidase X homolog; ClpX caseinolytic protease X homolog; energy-dependent regulator of proteolysis
Gene Aliases: AU014732; CLPX; E330029I21
UniProt ID: (Human) O76031, (Mouse) Q9JHS4, (Rat) Q5U2U0
Entrez Gene ID: (Human) 10845, (Mouse) 270166, (Rat) 300786
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support