Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MASNHKSSAA RPVSRGGVGL TGRPPSGIRP LSGNIRVATA MPPGTARPGS RGCPIGTGGV LSSQIKVAHR PVTQQGLTGM KTGTKGPQRQ ILDKSYYLGL LRSKISELTT EVNKLQKGIE MYNQENSVYL SYEKRAETLA VEIKELQGQL ADYNMLVDKL NTNTEMEEVM NDYNMLKAQN DRETQSLDVI FTERQAKEKQ IRSVEEEIEQ EKQATDDIIK NMSFENQVKY LEMKTTNEKL LQELDTLQQQ LDSQNMKKES LEAEIAHSQV KQEAVLLHEK LYELESHRDQ MIAEDKSIGS PMEEREKLLK QIKDDNQEIA SMERQLTDTK EKINQFIEEI RQLDMDLEEH QDPTNYGWKI LEKNTGSFKK QV; Positive Samples: 293T, A549, Mouse testis; Cellular Location: Cell projection, Cytoplasmic vesicle, cilium
This gene encodes a core intraflagellar transport (IFT) protein which belongs to a multi-protein complex involved in the transport of ciliary proteins along axonemal microtubules. IFT proteins are found at the base of the cilium as well as inside the cilium, where they assemble into long arrays between the ciliary base and tip. This protein, together with intraflagellar transport protein 81, binds and transports tubulin within cilia and is required for ciliogenesis. Naturally occurring mutations in this gene are associated with amyotrophic lateral sclerosis--frontotemporal dementia and Bardet-Biedl Syndrome.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Capillary morphogenesis gene 1 protein; capillary morphogenesis protein 1; CMG-1; coiled-coil domain containing 2; Coiled-coil domain-containing protein 2; intraflagellar transport 74 homolog; Intraflagellar transport protein 74 homolog
Gene Aliases: 1700029H06Rik; b2b796Clo; CCDC2; CMG-1; CMG1; IFT74
UniProt ID: (Human) Q96LB3, (Mouse) Q8BKE9
Entrez Gene ID: (Human) 80173, (Mouse) 67694
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support