Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the M20 metalloprotease family. The encoded protein is specifically expressed in the brain, is a homodimeric dipeptidase which was identified as human carnosinase. This gene contains trinucleotide (CTG) repeat length polymorphism in the coding region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-Ala-His dipeptidase; carnosinase 1; Carnosine dipeptidase 1; carnosine dipeptidase 1 (metallopeptidase M20 family); CNDP dipeptidase 1; Glutamate carboxypeptidase-like protein 2; Serum carnosinase
Gene Aliases: CN1; CNDP1; CPGL2; HsT2308; UNQ1915/PRO4380
UniProt ID: (Human) Q96KN2
Entrez Gene ID: (Human) 84735
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support