Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T, A431, Jurkat, Raji, mouse brain.
Predicted reactivity: Mouse (92%), Rat (92%), Zebrafish (81%), Xenopus laevis (80%), Pig (89%), Chicken (85%), Rhesus Monkey (93%), Chimpanzee (98%), Bovine (93%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CNDP2, also known as tissue carnosinase and peptidase A.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carnosine dipeptidase II; CNDP dipeptidase 2; Cytosolic non-specific dipeptidase; cytosolic nonspecific dipeptidase; epididymis secretory protein Li 13; Glutamate carboxypeptidase-like protein 1; peptidase 1; Peptidase A; Threonyl dipeptidase
Gene Aliases: 0610010E05Rik; C76600; CN2; CNDP2; CPGL; Dip-2; HEL-S-13; HsT2298; Pep-1; Pep1; PEPA
UniProt ID: (Human) Q96KP4, (Mouse) Q9D1A2
Entrez Gene ID: (Human) 55748, (Mouse) 66054
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support