Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VCISDNYWLG KKKPCITYGL RGICYFFIEV ECSNKDLHSG VYGGSVHEAM TDLILLMGSL VDKRGNILIP GINEAVAAVT EEEHKLYDDI DFDIEEFAKD VGAQILLHSH
CNDP2, also known as tissue carnosinase and peptidase A.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carnosine dipeptidase II; CNDP dipeptidase 2; Cytosolic non-specific dipeptidase; cytosolic nonspecific dipeptidase; epididymis secretory protein Li 13; Glutamate carboxypeptidase-like protein 1; Peptidase A; Threonyl dipeptidase
Gene Aliases: CN2; CNDP2; CPGL; HEL-S-13; HsT2298; PEPA
UniProt ID: (Human) Q96KP4
Entrez Gene ID: (Human) 55748
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support