Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MEPVTKWSPK QVVDWTRGLD DCLQQYVHKF EREKINGEQL LQISHQDLEE LGVTRIGHQE LVLEAVDLLC ALNYGLETDN MKNLVLKLRA SSHNLQNYIS SRRKSPAYDG NTSRKAPNEF LTSVVELIGA AKALLAWLDR APFTGITDFS VTKNKIIQLC LDLTTTVQKD CFVAEMEDKV LTVVKVLNGI CDKTIRSTTD PVMSQCACLE EVHLPNIKPG EGLGMYIKST YDGLHVITGT TENSPADRSQ KIHAGDEVIQ VNQQTVVGWQ LKNLVKKLRE NPTGVVLLLK KRPTGSFNFT PAPLKNLRWK PPLVQTSPPP ATTQSPESTM DTSLKKEKSA ILDLYIPPPP AVPYSPRDEN GSFVYGGSSK CKQPLPGPKG SESPNSFLDQ ESRRRRFTIA DSDQLPGYSV ETNILPTKMR EKTPSYGKPR PLSMPADGNW MGIVDPFARP RGHGRKGEDA LCRYFSNERI PPIIEESSSP PYRFSRPTTE RHLVRGADYI RGSRCYINSD LHSSATIPFQ EEGTKKKSGS SATKSSSTEP SLLVSWFTRL KLLTH
CNKSR3 may be involved in transepithelial sodium transport. Regulates aldosterone-induced and ENaC-mediated sodium transport possibly through regulation of the ERK pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CNK homolog protein 3; CNK3; CNKSR family member 3; Connector enhancer of kinase suppressor of ras 3; Connector enhancer of KSR 3; KOX14; Maguin-like protein; membrane associated guanylate kinase interacting protein-like 1; membrane associated guanylate kinase, WW and PDZ domain containing 1; membrane-associated guanylate kinase-interacting protein-like 1; ZNF21
Gene Aliases: CNKSR3; MAGI1
UniProt ID: (Human) Q6P9H4
Entrez Gene ID: (Human) 154043
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support