Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSSTQFNKGP SYGLSAEVKN RLLSKYDPQK EAELRTWIEG LTGLSIGPDF QKGLKDGTIL CTLMNKLQPG SVPKINRSMQ NWHQLENLSN FIKAMVSYGM NPVDLFEAND LFESGNMTQV QVSLLALAGK MGTNKCASQS GMTAYGTRRH LYDPKNHILP PMDHSTISLQ MGTNKCASQV GMTAPGTRRH IYDTKLGTDK CDNSSMSLQM GYTQGANQSG QVFGLGRQIY DPKYCPQGTV ADGAPSGTGD CPDPGEVPEY PPYYQEEAGY
The protein encoded by this gene, which can bind actin, calmodulin, troponin C, and tropomyosin, may function in the structural organization of actin filaments. The encoded protein could play a role in smooth muscle contraction and cell adhesion. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calponin H2, smooth muscle; Calponin-2; CLP2; Neutral calponin
Gene Aliases: CNN2
UniProt ID: (Human) Q99439
Entrez Gene ID: (Human) 1265
Molecular Function:
non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support