Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTHFNKGPS YGLSAEVKNK IASKYDHQAE EDLRNWIEEV TGMSIGPNFQ LGLKDGIILC ELINKLQPGS VKKVNESSLN WPQLENIGNF IKAIQAYGMK PHDIFEANDL FENGNMTQVQ TTLVALAGLA KTKGFHTTID IGVKYAEKQT RRFDEGKLKA GQSVIGLQMG TNKCASQAGM TAYGTRRHLY DPKMQTDKPF DQTTISLQMG TNKGASQAGM LAPGTRRDIY DQKLTLQPVD NSTISLQMGT NKVASQKGMS VYGLGRQVYD PKYCAAPTEP VIHNGSQGTG TNGSEISDSD YQAEYPDEYH GEYQDDYPRD YQYSDQGIDY (1-329 aa encoded by BC025372)
This gene encodes a protein with a markedly acidic C terminus; the basic N-terminus is highly homologous to the N-terminus of a related gene, CNN1. Members of the CNN gene family all contain similar tandemly repeated motifs. This encoded protein is associated with the cytoskeleton but is not involved in contraction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acidic calponin h3; calponin 3, acidic; Calponin, acidic isoform; Calponin, non-muscle isoform; Calponin-3; dJ639P13.2.2 (acidic calponin 3)
Gene Aliases: 1600014M03Rik; C85854; Calpo3; CNN3
UniProt ID: (Human) Q15417, (Mouse) Q9DAW9, (Rat) P37397
Entrez Gene ID: (Human) 1266, (Mouse) 71994, (Rat) 54321
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support